Skip to Content
Merck
N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine 99%

N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine

N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine 99%
Synonym(s):
PMDETA, PMDTA, N,N,N′,N′′,N′′-Pentamethyldiethylenetriamine
Linear Formula:
[(CH3)2NCH2CH2]2NCH3
CAS Number:
Molecular Weight:
173.30

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type