
N-Nitrosomethylphenylamine

Synonym(s):
N-Nitrosomethylphenylamine (NMPA), N-Methyl-N-nitrosoaniline, N-Methyl-N-nitrosobenzenamine, N-Methyl-N-phenylnitrosamine, NMPA
Empirical Formula (Hill Notation):
C7H8N2O
CAS Number:
Molecular Weight:
136.15
Products
Brand
Formula Weight
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing