Skip to Content
Merck
N,N′-Diphenylbenzidine 97%

N,N′-Diphenylbenzidine

N,N′-Diphenylbenzidine 97%
Synonym(s):
N,N′-Diphenylbenzidine
Linear Formula:
4-(C6H5NH)C6H4C6H4-4-(NHC6H5)
CAS Number:
Molecular Weight:
336.43

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing