Skip to Content
Merck
N,N′-Dimethylurea PESTANAL®, analytical standard

N,N′-Dimethylurea

N,N′-Dimethylurea PESTANAL®, analytical standard
Synonym(s):
1,3-Dimethylurea, N,N′-Dimethylurea
Linear Formula:
(CH3NH)2CO
CAS Number:
Molecular Weight:
88.11
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reaction Type
Reagent Type
Reference Material Type
Special Grade
Technique