N,N-Dimethylisobutyramide
Synonym(s):
N,N,2-Trimethylpropionamide
Linear Formula:
(CH3)2CHCON(CH3)2
CAS Number:
Molecular Weight:
115.17
Products
Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing