Skip to Content
Merck
N,N-Dimethylglycine ethyl ester 98%

N,N-Dimethylglycine ethyl ester

N,N-Dimethylglycine ethyl ester 98%
Linear Formula:
(CH3)2NCH2COOC2H5
CAS Number:
Molecular Weight:
131.17
EC Number:

Products

Shipping
Brand
Boiling Point (°C)
Formula Weight
Purity
Application
Markush Class
Markush Group
Amino Acid
Amino Acid Type
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing