Skip to Content
Merck
N-Methyl-N-trimethylsilyltrifluoroacetamide activated III for GC derivatization, LiChropur™, activated with imidazole

N-Methyl-N-trimethylsilyltrifluoroacetamide activated III

N-Methyl-N-trimethylsilyltrifluoroacetamide activated III for GC derivatization, LiChropur™, activated with imidazole
Synonym(s):
N-Methyl-N-trimethylsilyltrifluoroacetamide imidazole mixture, MSTFA activated III
CAS Number:

Products

Brand
Quality Segment
Reaction Type
Reagent Type
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing