
Nitinol-core (NIT) SPME Fiber Assembly

Synonym(s):
SPME fiber assembly Carboxen/Polydimethylsiloxane (CAR/PDMS), SPME Fiber Assembly
Products
Shipping
Brand
Sustainability
Feature
Length (cm)
Color
df (µm)
Package Size
Size
Gauge
Technique
Showing 1-8 of 8
Compare
Product Number
Description
Pricing