Nicotine Related Compound A/EP Impurity A
Synonym(s):
Anatabine, (RS)-Anatabine, (±)-Anatabine, 1,2,3,6-Tetrahydro-2,3′-bipyridine, 1,2,3,6-Tetrahydro-2,3′-dipyridine
Empirical Formula (Hill Notation):
C10H12N2
CAS Number:
Molecular Weight:
160.22
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing