![Neotame Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/401/761/f42e50b1-e744-48ff-8034-5ff69389efdf/640/f42e50b1-e744-48ff-8034-5ff69389efdf.jpg)
Neotame
![Neotame Pharmaceutical Secondary Standard; Certified Reference Material](/deepweb/assets/sigmaaldrich/product/images/401/761/f42e50b1-e744-48ff-8034-5ff69389efdf/640/f42e50b1-e744-48ff-8034-5ff69389efdf.jpg)
Synonym(s):
Neotame monohydrate, N-[N-(3,3-Dimethylbutyl)-L-α-aspartyl]-L-phenylalanine 1-methyl ester monohydrate
Empirical Formula (Hill Notation):
C20H30N2O5 · H2O
CAS Number:
Molecular Weight:
396.48
Products
Shipping
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing