Skip to Content
Merck
Neocuproine ≥98%

Neocuproine

Neocuproine ≥98%
Synonym(s):
2,9-Dimethyl-1,10-phenanthroline, DMPHEN, Neocuproine
Empirical Formula (Hill Notation):
C14H12N2
CAS Number:
Molecular Weight:
208.26

Products

Shipping
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Analyte Suitability
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing