![Neoandrographolide analytical standard](/deepweb/assets/sigmaaldrich/product/structures/152/028/fbeb26b8-1135-4a93-b6a8-63ed3c5b8933/640/fbeb26b8-1135-4a93-b6a8-63ed3c5b8933.png)
Neoandrographolide
![Neoandrographolide analytical standard](/deepweb/assets/sigmaaldrich/product/structures/152/028/fbeb26b8-1135-4a93-b6a8-63ed3c5b8933/640/fbeb26b8-1135-4a93-b6a8-63ed3c5b8933.png)
Synonym(s):
ent-19-Hydroxy-8(17),13-labdadien-16,15-olide 19-O-β-D-glucopyranoside, D-Glucopyranosyloxy)methyl]decahydro-5,8a-dimethyl-2-methylene-1-naphthalenyl]ethyl]-2(5H)-furanone
Empirical Formula (Hill Notation):
C26H40O8
CAS Number:
Molecular Weight:
480.59
Products
Shipping
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing