Skip to Content
Merck
N-Cyclopropyl-2,4,6-triamino-1,3,5-triazine 97%

N-Cyclopropyl-2,4,6-triamino-1,3,5-triazine

N-Cyclopropyl-2,4,6-triamino-1,3,5-triazine 97%
Synonym(s):
N-Cyclopropyl-1,3,5-triazin-2,4,6-triamine, Cyromazin
Empirical Formula (Hill Notation):
C6H10N6
CAS Number:
Molecular Weight:
166.18
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing