Skip to Content
Merck
N-Benzylidenebenzenesulfonamide 97%

N-Benzylidenebenzenesulfonamide

N-Benzylidenebenzenesulfonamide 97%
Linear Formula:
C6H5SO2N=CHC6H5
CAS Number:
Molecular Weight:
245.30

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing