N-Acetylprocainamide hydrochloride
Synonym(s):
N-Acetylnovocainamide hydrochloride, Acecainide hydrochloride, NAPA
Linear Formula:
4-(CH3CONH)C6H4CONHCH2CH2N(C2H5)2·HCl
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing