Skip to Content
Merck
N-Acetyl-L-aspartic acid ≥99.0% (T)

N-Acetyl-L-aspartic acid

N-Acetyl-L-aspartic acid ≥99.0% (T)
Linear Formula:
HO2CCH2CH(NHCOCH3)CO2H
CAS Number:
Molecular Weight:
175.14

Products

Shipping
Product Category
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Amino Acid Type
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing