Skip to Content
Merck
Mycophenolate Mofetil Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material, pkg of 100 mg

Mycophenolate Mofetil Related Compound B

Mycophenolate Mofetil Related Compound B Pharmaceutical Secondary Standard; Certified Reference Material, pkg of 100 mg
Synonym(s):
(RS)-7-Hydroxy-5-methoxy-4-methyl-6-[2-(2-methyl-5-oxo-tetrahydrofuran-2-yl)ethyl]-3H-isobenzofuran-1-one, Mycophenolic acid lactone
Empirical Formula (Hill Notation):
C17H20O6
CAS Number:
Molecular Weight:
320.34

Products

Shipping
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade