Skip to Content
Merck
vibrant-m

Moxifloxacin Related Compound G

vibrant-m
Synonym(s):
1-Cyclopropyl-6-fluoro-1,4-dihydro-8-methoxy-7-[(4aR,7aR)-octahydro-6H-pyrrolo[3,4-b]pyridin-6-yl]-4-oxo-3-quinolinecarboxylic acid monohydrochloride
Empirical Formula (Hill Notation):
C21H24FN3O4 · HCl
CAS Number:
Molecular Weight:
437.89

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1