![Moxifloxacin hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/181/466/f118f724-9602-48a6-b896-b7654ebd4412/640/f118f724-9602-48a6-b896-b7654ebd4412.png)
Moxifloxacin hydrochloride
![Moxifloxacin hydrochloride European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/181/466/f118f724-9602-48a6-b896-b7654ebd4412/640/f118f724-9602-48a6-b896-b7654ebd4412.png)
Synonym(s):
1-Cyclopropyl-6-fluoro-8-methoxy-7-[(4aS,7aS)-octahydropyrrolo[3,4-b]pyridin-6-yl]-4-oxo-1,4-dihydroquinoline-3-carboxylic acid hydrochloride, Moxifloxacin hydrochloride
Empirical Formula (Hill Notation):
C21H24FN3O4· HCl
CAS Number:
Molecular Weight:
437.89
Products
Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Technique
Showing 1-5 of 5
Compare
Product Number
Description
Pricing