Monoclonal Anti-MAP2 antibody produced in mouse
Synonym(s):
MAP2A, MAP2B, MAP2C
Products
Product Category
Brand
Biological Source
Clonality
Gene Alias
Isotype
Physical Form
Product Line
Antibody Form
Research Area
Conjugate
Quality Segment
Package Size
Post Translational Modification
Species Reactivity
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing