![Methyl-p-benzoquinone analytical standard](/deepweb/assets/sigmaaldrich/product/structures/149/953/f4b206e2-44bc-4da4-ba9b-fe176eb24ec0/640/f4b206e2-44bc-4da4-ba9b-fe176eb24ec0.png)
Methyl-p-benzoquinone
![Methyl-p-benzoquinone analytical standard](/deepweb/assets/sigmaaldrich/product/structures/149/953/f4b206e2-44bc-4da4-ba9b-fe176eb24ec0/640/f4b206e2-44bc-4da4-ba9b-fe176eb24ec0.png)
Products
Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Format
Quality Segment
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing