Skip to Content
Merck
Methyl benzoylformate 98%

Methyl benzoylformate

Methyl benzoylformate 98%
Synonym(s):
Methyl phenylglyoxylate, Methyl 2-oxo-2-phenylacetate
Linear Formula:
C6H5COCOOCH3
CAS Number:
Molecular Weight:
164.16

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
API Family
Special Grade