Skip to Content
Merck
Methyl 2-oxo-2H-pyran-3-carboxylate 98%

Methyl 2-oxo-2H-pyran-3-carboxylate

Methyl 2-oxo-2H-pyran-3-carboxylate 98%
Synonym(s):
Methyl 2-pyrone-3-carboxylate
Empirical Formula (Hill Notation):
C7H6O4
CAS Number:
Molecular Weight:
154.12
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing