![Methoxypolyethylene glycol amine 20,000](/deepweb/assets/sigmaaldrich/product/structures/266/027/c26b38a3-078d-4a3f-b9ca-7609fb1a1ede/640/c26b38a3-078d-4a3f-b9ca-7609fb1a1ede.png)
Methoxypolyethylene glycol amine
![Methoxypolyethylene glycol amine 20,000](/deepweb/assets/sigmaaldrich/product/structures/266/027/c26b38a3-078d-4a3f-b9ca-7609fb1a1ede/640/c26b38a3-078d-4a3f-b9ca-7609fb1a1ede.png)
Synonym(s):
O-(2-Aminoethyl)-O′-methylpolyethylene glycol, Aminopolyethylene glycol monomethyl ether, Methoxypolyoxyethylene amine
Linear Formula:
H2NCH2CH2(OCH2CH2)nOCH3
CAS Number:
Products
Shipping
Brand
Color
Physical Form
Quality Segment
Showing 1-5 of 5
Compare
Product Number
Description
Pricing