Skip to Content
Merck
Methoxsalen pharmaceutical secondary standard, certified reference material

Methoxsalen

Methoxsalen pharmaceutical secondary standard, certified reference material
Synonym(s):
8-Methoxypsoralen, 8-MOP, 9-Methoxyfuro[3,2-g][1]benzopyran-7-one, Ammoidin, Methoxsalen, Xanthotoxin
Empirical Formula (Hill Notation):
C12H8O4
CAS Number:
Molecular Weight:
216.19

Products

Shipping
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade