
Methacholine chloride

Synonym(s):
Acetyl-β-methylcholine chloride, (2-Acetoxypropyl)trimethylammonium chloride, Methacholine chloride
Linear Formula:
CH3COOCH(CH3)CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
195.69
Products
Shipping
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade
Showing 1-2 of 2