Skip to Content
Merck
Metaxalone pharmaceutical secondary standard, certified reference material

Metaxalone

Metaxalone pharmaceutical secondary standard, certified reference material
Synonym(s):
5-(3,5-Dimethylphenoxymethyl)-2-oxazolidinone, 5-[(3,5-Xylyloxy)methyl]-2-oxazolidinone, AHR 438, Metaxalon, Metazalone, Metazolone, NSC 170959, Metaxalone
Empirical Formula (Hill Notation):
C12H15NO3
CAS Number:
Molecular Weight:
221.25

Products

Shipping
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade