![MCPA TraceCERT®, certified reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/549/907/41a5ce0c-5862-4935-a2af-a1b82eeb2d04/640/41a5ce0c-5862-4935-a2af-a1b82eeb2d04.png)
MCPA
![MCPA TraceCERT®, certified reference material, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/549/907/41a5ce0c-5862-4935-a2af-a1b82eeb2d04/640/41a5ce0c-5862-4935-a2af-a1b82eeb2d04.png)
Synonym(s):
4-Chloro-2-methylphenoxyacetic acid, 4-Chloro-o-tolyloxyacetic acid, MCPA
Linear Formula:
ClC6H3(CH3)OCH2CO2H
CAS Number:
Molecular Weight:
200.62
Products
Shipping
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing