Skip to Content
Merck
Manganese(II) nitrate tetrahydrate purum p.a., ≥97.0% (KT)

Manganese(II) nitrate tetrahydrate

Manganese(II) nitrate tetrahydrate purum p.a., ≥97.0% (KT)
Synonym(s):
Manganese(II) nitrate tetrahydrate
Linear Formula:
Mn(NO3)2 · 4 H2O
CAS Number:
Molecular Weight:
251.01

Products

Shipping
Product Category
Brand
Sustainability
Color
Feature
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Special Grade