Skip to Content
Merck
Magnesium nitrate hexahydrate BioUltra, ≥99.0% (KT)

Magnesium nitrate hexahydrate

Magnesium nitrate hexahydrate BioUltra, ≥99.0% (KT)
Synonym(s):
Magnesium nitrate hexahydrate
Linear Formula:
Mg(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
256.41
EC Number:

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade