Skip to Content
Merck
Magnesium ionophore I Selectophore™, function tested

Magnesium ionophore I

Magnesium ionophore I Selectophore™, function tested
Synonym(s):
ETH 1117, Magnesium-ligand, N,N′-Diheptyl-N,N′-dimethyl-1,4-butanediamide
Empirical Formula (Hill Notation):
C20H40N2O2
CAS Number:
Molecular Weight:
340.54

Products

Shipping
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing