Skip to Content
Merck
Magnesium acetate tetrahydrate ≥99.9% trace metals basis

Magnesium acetate tetrahydrate

Magnesium acetate tetrahydrate ≥99.9% trace metals basis
Synonym(s):
Acetic acid magnesium salt, Magnesium acetate tetrahydrate
Linear Formula:
(CH3COO)2Mg · 4H2O
CAS Number:
Molecular Weight:
214.45

Products

Shipping
Product Category
Brand
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Special Grade