Skip to Content
Merck
L-Tyrosine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

L-Tyrosine

L-Tyrosine certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Synonym(s):
(S)-2-Amino-3-(4-hydroxyphenyl)propionic acid, 3-(4-Hydroxyphenyl)-L-alanine, L-Tyrosine
Linear Formula:
4-(HO)C6H4CH2CH(NH2)CO2H
CAS Number:
Molecular Weight:
181.19
EC Number:

Products

Shipping
Product Category
Brand
Biological Source
Color
Feature
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Analyte Suitability
Quality Segment
Application
Optical Isomer
Amino Acid
API Family
Reference Material Type
Special Grade
Technique