Skip to Content
Merck
L-Tyrosine-13C9,15N endotoxin tested, 98 atom % 13C, 98 atom % 15N, 98% (CP)

L-Tyrosine-13C9,15N

L-Tyrosine-13C9,15N endotoxin tested, 98 atom % 13C, 98 atom % 15N, 98% (CP)
Synonym(s):
L-4-Hydroxyphenylalanine-13C9,15N
Linear Formula:
4-HO-13C6H413CH213CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
191.12

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique