L-(−)-Sorbose
Synonym(s):
L(-)Sorbose
Empirical Formula (Hill Notation):
C6H12O6
CAS Number:
Molecular Weight:
180.16
Products
Shipping
Product Category
Brand
Color
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Optical Isomer
Special Grade
Showing 1-2 of 2