L-Phenylalanine-13C9,15N
Synonym(s):
Phenylalanine-UL-13C,15N
Linear Formula:
13C6H513CH213CH(15NH2)13CO2H
CAS Number:
Molecular Weight:
175.12
Products
Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-3 of 3
Compare
Product Number
Description
Pricing