Skip to Content
Merck
vibrant-m

Lopinavir for peak identification

vibrant-m
Synonym(s):
Lopinavir
Empirical Formula (Hill Notation):
C37H48N4O5
CAS Number:
Molecular Weight:
628.80

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing