Skip to Content
Merck
L-Leucine-13C6 endotoxin tested, ≥99 atom % 13C, ≥97% (CP)

L-Leucine-13C6

L-Leucine-13C6 endotoxin tested, ≥99 atom % 13C, ≥97% (CP)
Synonym(s):
(S)-2-Amino-4-methylpentanoic acid-13C6, 13C Labeled L-leucine
Linear Formula:
(13CH3)213CH13CH213CH(NH2)13CO2H
CAS Number:
Molecular Weight:
137.13

Products

Shipping
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Optical Isomer
Technique
Showing 1-2 of 2