Skip to Content
Merck
Lithium tetrafluoroborate Technipur®, ultra dry, powder, 99.99% trace metals basis

Lithium tetrafluoroborate

Lithium tetrafluoroborate Technipur®, ultra dry, powder, 99.99% trace metals basis
Synonym(s):
Lithium borofluoride, Lithium fluoroborate, Lithium tetrafluoroborate
Linear Formula:
LiBF4
CAS Number:
Molecular Weight:
93.75

Products

Shipping
Product Category
Brand
Sustainability
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Special Grade