Lithium carbamoylphosphate dibasic hydrate
Synonym(s):
CAP-Li2, Carbamoylphosphate dilithium salt, Carbamyl phosphate dilithium salt, Carbamyl phosphoric acid dilithium salt
Linear Formula:
NH2COOPO3Li2·xH2O
CAS Number:
Molecular Weight:
152.89 (anhydrous basis)
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Showing 1-1 of 1
Compare
Product Number
Description
Pricing