Skip to Content
Merck
vibrant-m

Levomepromazine for system suitability

vibrant-m
Synonym(s):
Methotrimeprazine, (−)-10-(3-[Dimethylamino]-2-methylpropyl)-2-methoxyphenothiazine, Levomeprazine
Empirical Formula (Hill Notation):
C19H24N2OS
CAS Number:
Molecular Weight:
328.47

Products

Formula Weight
Manufacturer
Application
Special Grade
Showing 1-1 of 1