L-(+)-2-Amino-4-phosphonobutyric acid
Synonym(s):
L-AP-4, L-AP4
Empirical Formula (Hill Notation):
C4H10NO5P
CAS Number:
Molecular Weight:
183.10
Products
Shipping
Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing