
Lauric acid-1-13C

Synonym(s):
Dodecanoic acid-1-13C
Linear Formula:
CH3(CH2)1013CO2H
CAS Number:
Molecular Weight:
201.31
Products
Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing