Skip to Content
Merck
vibrant-m

Latanoprost Related Compound A

vibrant-m
Synonym(s):
Isopropyl (E)-7-[(1R,2R,3R,5S)-3,5-dihydroxy-2-[(3R)-3-hydroxy-5-phenylpentyl]cyclopentyl]hept-5-enoate, 5-trans-Latanoprost
Empirical Formula (Hill Notation):
C26H40O5
CAS Number:
Molecular Weight:
432.59

Products

Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1