![Larixyl acetate phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/274/978/8da67647-42e3-4b28-8376-6e97a50521bc/640/8da67647-42e3-4b28-8376-6e97a50521bc.png)
Larixyl acetate
![Larixyl acetate phyproof® Reference Substance](/deepweb/assets/sigmaaldrich/product/structures/274/978/8da67647-42e3-4b28-8376-6e97a50521bc/640/8da67647-42e3-4b28-8376-6e97a50521bc.png)
Synonym(s):
(αS,1S,4S,4aS,8aR)-4-(Acetyloxy)-α-ethenyldecahydro-α,5,5,8a-tetramethyl-2-methylene-1-naphthalenepropanol, Larixol acetate
Empirical Formula (Hill Notation):
C22H36O3
CAS Number:
Molecular Weight:
348.52
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing