Skip to Content
Merck
Lamotrigine British Pharmacopoeia (BP) Reference Standard

Lamotrigine

Lamotrigine British Pharmacopoeia (BP) Reference Standard
Synonym(s):
6-(2,3-Dichlorophenyl)-1,2,4-triazine-3,5-diamine, GI 267119X, Lamotrigine
Empirical Formula (Hill Notation):
C9H7Cl2N5
CAS Number:
Molecular Weight:
256.09
EC Number:

Products

Shipping
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Agency
Format
Research Area
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique