Skip to Content
Merck
KT 5720 ≥98% (HPLC), powder

KT5720

KT 5720 ≥98% (HPLC), powder
Synonym(s):
KT5720, (9S,10S,12R)-2,3,9,10,11,12-Hexahydro-10-hydroxy-9-methyl-1-oxo-9,12-epoxy-1H-diindolo[1,2,3-fg:3′,2′,1′-kl]pyrrolo[3,4-i][1,6]benzodiazocine-10-carboxylic acid hexyl ester, InSolution KT5720
Empirical Formula (Hill Notation):
C32H31N3O5
CAS Number:
Molecular Weight:
537.61

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Research Area
Quality Segment
Showing 1-3 of 3