Isoverbascoside
Synonym(s):
Isoacteoside, (E)-2-(3,4-dihydroxyphenyl)ethyl 3-O-(6-deoxy-α-L-mannopyranosyl)-, 6-(3-(3,4-dihydroxyphenyl)-β-D-glucopyranoside, Isoverbascoside
Empirical Formula (Hill Notation):
C29H36O15
CAS Number:
Molecular Weight:
624.59
Products
Shipping
Formula Weight
Manufacturer
Product Line
Purity
Format
Application
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing