Skip to Content
Merck
Isoproterenol Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material

Isoproterenol hydrochloride

Isoproterenol Hydrochloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Isoprenaline hydrochloride, (±)-Isoproterenol hydrochloride, 1-(3′,4′-Dihydroxyphenyl)-2-isopropylaminoethanol hydrochloride, N-Isopropyl-DL-noradrenaline hydrochloride
Empirical Formula (Hill Notation):
C11H17NO3 · HCl
CAS Number:
Molecular Weight:
247.72

Products

Shipping
Brand
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade