Skip to Content
Merck
Isatoic anhydride 96%

Isatoic anhydride

Isatoic anhydride 96%
Synonym(s):
3,1-Benzoxazine-2,4(1H)-dione, Anthranilic acid N-carboxylic acid anhydride, Isatoic anhydride
Empirical Formula (Hill Notation):
C8H5NO3
CAS Number:
Molecular Weight:
163.13

Products

Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing